missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Tryptophan hydroxylase 2 Polyclonal antibody specifically detects Tryptophan hydroxylase 2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Tryptophan hydroxylase 2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ADHD7, EC 1.14.16, EC 1.14.16.4, FLJ37295, MGC138872, Neuronal tryptophan hydroxylase, NTPHMGC138871, tryptophan 5-hydroxylase 2, Tryptophan 5-monooxygenase 2, tryptophan hydroxylase 2 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide made to an N-terminal portion of the human Tryptophan hydroxylase 2 protein (between residues 1-100) [Uniprot: Q8IWU9] |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?