Learn More
Tryptophan hydroxylase 2 Antibody (CL2990) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46646F
Product Code. 30303760
  • £558.00 / 0.10mL
Description
Tryptophan hydroxylase 2 Monoclonal antibody specifically detects Tryptophan hydroxylase 2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tryptophan hydroxylase 2
Monoclonal
FITC
ADHD7, EC 1.14.16, EC 1.14.16.4, FLJ37295, MGC138872, Neuronal tryptophan hydroxylase, NTPHMGC138871, tryptophan 5-hydroxylase 2, Tryptophan 5-monooxygenase 2, tryptophan hydroxylase 2
This antibody was developed using a recombinant protein derived from Q8IWU9, with the exact immunogen sequence remaining proprietary.
0.1 mL
Lipid and Metabolism, Phospho Specific
121278
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL2990
PBS
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified