missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophan hydroxylase 2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £310.00
Specifications
| Antigen | Tryptophan hydroxylase 2 |
|---|---|
| Concentration | 1.19 mg/mL |
| Dilution | Western Blot 2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18260796
|
Novus Biologicals
NBP1-77556SS |
0.025 mL |
£150.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18209735
|
Novus Biologicals
NBP1-77556 |
0.1 mL |
£310.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tryptophan hydroxylase 2 Polyclonal specifically detects Tryptophan hydroxylase 2 in Human samples. It is validated for Western Blot.Specifications
| Tryptophan hydroxylase 2 | |
| Western Blot 2 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| ADHD7, EC 1.14.16, EC 1.14.16.4, FLJ37295, MGC138872, Neuronal tryptophan hydroxylase, NTPHMGC138871, tryptophan 5-hydroxylase 2, Tryptophan 5-monooxygenase 2, tryptophan hydroxylase 2 | |
| TPH2 | |
| IgG | |
| Affinity Purified |
| 1.19 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Q8IWU9 | |
| 121278 | |
| A synthetic peptide made to an N-terminal portion of the human Tryptophan hydroxylase 2 protein (between residues 1-100) [Uniprot: Q8IWU9] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title