missing translation for 'onlineSavingsMsg'
Learn More

Tryptophan hydroxylase 2 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18373421 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373421 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373421 Supplier Novus Biologicals Supplier No. NBP177559AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Tryptophan hydroxylase 2 Polyclonal specifically detects Tryptophan hydroxylase 2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tryptophan hydroxylase 2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ADHD7, EC 1.14.16, EC 1.14.16.4, FLJ37295, MGC138872, Neuronal tryptophan hydroxylase, NTPHMGC138871, tryptophan 5-hydroxylase 2, Tryptophan 5-monooxygenase 2, tryptophan hydroxylase 2
Gene Symbols TPH2
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Tryptophan hydroxylase 2 protein (between residues 100-200) [UniProtNo. Q8IWU9]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 121278
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.