missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophan Hydroxylase 1/TPH-1 Rabbit anti-Human, Clone: 145, Novus Biologicals™
Rabbit Monoclonal Antibody
£296.00 - £429.00
Specifications
| Antigen | Tryptophan Hydroxylase 1/TPH-1 |
|---|---|
| Clone | 145 |
| Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 ul/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261648
|
Novus Biologicals
NBP2-90022-50UL |
50 μg |
£296.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18284616
|
Novus Biologicals
NBP2-90022-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tryptophan Hydroxylase 1/TPH-1 Monoclonal specifically detects Tryptophan Hydroxylase 1/TPH-1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Tryptophan Hydroxylase 1/TPH-1 | |
| Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 ul/mg of lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.14.16, EC 1.14.16.4, indoleacetic acid-5-hydroxylase, L-tryptophan hydroxylase, MGC119994, TPH, TPRH, TRPH, tryptophan 5-hydroxylase 1, Tryptophan 5-monooxygenase 1, tryptophan hydroxylase (tryptophan 5-monooxygenase), tryptophan hydroxylase 1 | |
| Recombinant Human Tryptophan Hydroxylase 1/TPH-1 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 145 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism, Neuroscience, Neurotransmission, Phospho Specific | |
| 0.2 um filtered solution in PBS | |
| 7166 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title