missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Tryptophan Hydroxylase 1/TPH-1 Antibody (TPH1/7662), Novus Biologicals™
Mouse Monoclonal Antibody
£178.00 - £351.00
Specifications
| Antigen | Tryptophan Hydroxylase 1/TPH-1 |
|---|---|
| Clone | TPH1/7662 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043201
|
Novus Biologicals™
NBP3-26737-100ug |
100 μg |
£351.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043400
|
Novus Biologicals™
NBP3-26737-20ug |
20 μg |
£178.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tryptophan Hydroxylase 1/TPH-1 Monoclonal antibody specifically detects Tryptophan Hydroxylase 1/TPH-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Tryptophan Hydroxylase 1/TPH-1 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism, Neuroscience, Neurotransmission, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 7166 | |
| IgG2 κ | |
| Protein A or G purified |
| TPH1/7662 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.14.16, EC 1.14.16.4, indoleacetic acid-5-hydroxylase, L-tryptophan hydroxylase, MGC119994, TPH, TPRH, TRPH, tryptophan 5-hydroxylase 1, Tryptophan 5-monooxygenase 1, tryptophan hydroxylase (tryptophan 5-monooxygenase), tryptophan hydroxylase 1 | |
| Recombinant full-length human Tryptophan Hydroxylase 1/TPH-1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title