missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Soy Broth (Soybean Casein Digest Medium EP/USP/BP/JP) (Dehydrated)

Product Code. 10535943
Change view
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
Unit Size:
2.5kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10535943 5 kg 5kg
11409658 2.5 kg 2.5kg
10525943 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10535943 Supplier Thermo Scientific™ Supplier No. CM0129T
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Grow a wide variety of microorganisms with this general purpose medium containing two peptones.

Grow a wide variety of microorganisms with Thermo Scientific™ Oxoid™ Tryptone Soya Broth EP/USP/BP/JP (Dehydrated). The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4 and food testing methods for the growth of a wide variety of organisms.

Use Tryptone Soya Broth to support the luxuriant growth of many fastidious organisms, without the addition of serum, while conforming to the formulations detailed in various International Pharmacopoeias.

  • Suitable for tube dilution method of antibiotic susceptibility testing
  • Recommended in the USP for the recovery of organisms after sterilization processes4

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Tryptone Soy Broth (Soybean Casein Digest Medium USP)
Form Powder
Media Type Tryptone Soy Broth
Product Type Nutrient Broth
Quantity 5 kg
Yield For 167.0L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.