missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Tryptone Large Granules

Product Code. 12861660
Change view
Click to view available options
Quantity:
500 g
2000 g
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12861660 500 g 500g
12851660 2000 g 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12861660 Supplier Fisher BioReagents™ Supplier No. 12861660
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Used as a nitrogen source in culture media for bacteria and fungi.

  • Pancreatic digest of Casein
  • Light beige
  • Particle size: 1 to 2mm

Specifications

Product Type Dehydrated Culture Media
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.