missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tryptase Monoclonal Antibody (C13), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£161.00 - £389.00

Specifications

Antigen Tryptase
Clone C13
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17939732
GREENER_CHOICE
View Documents
Invitrogen™
MA551031
200 μL
£389.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17900063
View Documents
Invitrogen™
MA551032
20 μL
£161.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tryptase is a protein that belongs to the family of serine proteases and is primarily found in mast cells. It plays a crucial role in the immune response by cleaving and activating other proteins involved in inflammation and tissue repair. Tryptase is also involved in the regulation of blood pressure and has been implicated in various diseases, including asthma, allergies, and cardiovascular disorders. Its structure consists of four identical subunits that form a tetramer, and it is activated by proteolytic cleavage of its proenzyme form.
Specifications

Specifications

Tryptase
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
TPSG1/TPSB2/TPSAB1/TPSD1
MCT7
IgG2b κ
Protein A/G
MCT7
C13
Western Blot
Unconjugated
Mouse
Canine, Pig
Q9N2D1
397389
Recombinant Dog TPS protein, Thr151-Pro275 (Accession #P15944).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.