missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptase gamma-1/TPSG1 Antibody (OTI1E11), DyLight 594, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Tryptase gamma-1/TPSG1 Monoclonal antibody specifically detects Tryptase gamma-1/TPSG1 in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
Specifications
| Antigen | Tryptase gamma-1/TPSG1 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI1E11 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.4.21, EC 3.4.21.-, gamma I, gamma II, lung tryptase, mast cell protease II, mast cell tryptase, PRSS31skin tryptase, Serine protease 31, TMTpituitary tryptase, Transmembrane tryptase, trpA, tryptase gamma, tryptase gamma 1, tryptase gamma I, tryptase gamma II |
| Host Species | Mouse |
| Immunogen | Recombinant protein fragment corresponding to amino acids 20-283 of human Tryptase gamma-1/TPSG1 (NP_036599) produced in E.coli. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?