missing translation for 'onlineSavingsMsg'
Learn More

Tryptase gamma-1/TPSG1 Antibody (279005) [HRP], Novus Biologicals™

Product Code. 30732784 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732784 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732784 Supplier Novus Biologicals Supplier No. FAB1667H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Tryptase gamma-1/TPSG1 Monoclonal antibody specifically detects Tryptase gamma-1/TPSG1 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tryptase gamma-1/TPSG1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 279005
Conjugate HRP
Formulation PBS
Gene Alias EC 3.4.21, EC 3.4.21.-, gamma I, gamma II, lung tryptase, mast cell protease II, mast cell tryptase, PRSS31skin tryptase, Serine protease 31, TMTpituitary tryptase, Transmembrane tryptase, trpA, tryptase gamma, tryptase gamma 1, tryptase gamma I, tryptase gamma II
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Tryptase gamma -1/TPSG1, Arg20-Arg281, Accession # Q9NRR2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25823
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.