missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptase beta-2/TPSB2 Antibody (9H6), Novus Biologicals™
Rabbit Monoclonal Antibody
£269.00 - £431.00
Specifications
| Antigen | Tryptase beta-2/TPSB2 |
|---|---|
| Clone | 9H6 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30025108
|
Novus Biologicals
NBP3-26273-50ul |
50 μL |
£269.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30038751
|
Novus Biologicals
NBP3-26273-100ul |
100 μL |
£431.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tryptase beta-2/TPSB2 Monoclonal antibody specifically detects Tryptase beta-2/TPSB2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
| Tryptase beta-2/TPSB2 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 3.4.21, EC 3.4.21.59, mast cell tryptase beta II, mast cell tryptase beta III, TPS2tryptase beta-2, tryptase beta 2, tryptase beta 2 (gene/pseudogene), tryptase beta II, tryptase beta III, Tryptase II, tryptase III, tryptase-2, tryptaseB, tryptaseC | |
| A synthesized peptide derived from Human Tryptase beta-2/TPSB2 [UniProt P20231] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 9H6 | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Immunology, Innate Immunity | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 64499 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title