missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptase beta-2/TPSB2 Antibody (03), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06181-100ul
This item is not returnable.
View return policy
Description
Tryptase beta-2/TPSB2 Monoclonal antibody specifically detects Tryptase beta-2/TPSB2 in Human samples. It is validated for ELISA
Specifications
| Tryptase beta-2/TPSB2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 03 | |
| ELISA 1:1000-1:2000 | |
| EC 3.4.21, EC 3.4.21.59, mast cell tryptase beta II, mast cell tryptase beta III, TPS2tryptase beta-2, tryptase beta 2, tryptase beta 2 (gene/pseudogene), tryptase beta II, tryptase beta III, Tryptase II, tryptase III, tryptase-2, tryptaseB, tryptaseC | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Tryptase beta-2/TPSB2 (Accession#: NP_077078.5; Met 1-Pro 275). | |
| 100 μg | |
| Cell Biology, Immunology, Innate Immunity | |
| 64499 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction