missing translation for 'onlineSavingsMsg'
Learn More

Tryptamine, TRC

Product Code. 17982135
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982135 10 g 10g
17931825 50 g 50g
17962355 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17982135 Supplier Toronto Research Chemicals Supplier No. TRCT89460010G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-54-1
Molecular Formula C10 H12 N2
Molecular Weight (g/mol) 160.22
Synonym Indole, 3-(2-aminoethyl)- (8CI), 2-(1H-Indol-3-yl)ethan-1-amine, 2-(1H-Indol-3-yl)ethanamine, 2-(1H-Indol-3-yl)ethylamine, 2-(3-Indolyl)ethylamine, 3-(2-Aminoethyl)-1H-indole, 3-(2-Aminoethyl)indole, 3-Indoleethanamine, 3-Indoleethylamine, NSC 165212, Tryptamin, Tryptamine, β-(3-Indolyl)ethylamine, 1H-Indole-3-ethanamine
IUPAC Name 2-(1H-indol-3-yl)ethanamine
SMILES NCCc1c[nH]c2ccccc12

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H12N2/c11-6-5-8-7-12-10-4-2-1-3-9(8)10/h1-4,7,12H,5-6,11H2
Formula Weight 160.1
Percent Purity >95%
Chemical Name or Material Tryptamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.