missing translation for 'onlineSavingsMsg'
Learn More

Trypsinogen Antibody, HRP, Novus Biologicals™

Product Code. 18357320 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357320 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18357320 Supplier Novus Biologicals Supplier No. NBP17769750UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Trypsinogen Polyclonal specifically detects Trypsinogen in Bovine samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trypsinogen
Applications ELISA, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate HRP
Dilution Western Blot 1:1000 - 1:5000, ELISA 1:10000 - 1:40000
Formulation 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2, 10Mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate
Gene Alias 2, Brain trypsinogen, Cationic Trypsinogen, Mesotrypsinogen, Pancreatic anionic trypsinogen, Protease, PRSS2, PRSS3, PRSS4, serine, TRP 1, TRP1, TRY1, TRY2, TRY3. TRY4, TRYP1, TRYP2, TRYP8, Trypsin 2, Trypsin 3, Trypsin III, Trypsin IV
Host Species Rabbit
Immunogen Trypsinogen [Bovine Pancreas]. Uniprot ID Q29463
Purification Method Delipidation and Defibrination
Quantity 50 μg
Primary or Secondary Primary
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum as well as purified and partially purified Trypsinogen from bovine pancreas.
Reconstitution Reconstitute with 100μL deionized water.
Target Species Bovine
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.