missing translation for 'onlineSavingsMsg'
Learn More

Trypsin Inhibitor Antibody, Novus Biologicals™

Product Code. 18340900 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340900 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340900 Supplier Novus Biologicals Supplier No. NBP1780880.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Trypsin Inhibitor Polyclonal specifically detects Trypsin Inhibitor in Plant samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trypsin Inhibitor
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:3000, ELISA 1:5000 - 1:25000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Alias KTI3, Kunitz trypsin inhibitor
Gene Symbols KTI3
Host Species Rabbit
Immunogen Trypsin Inhibitor [Lima Bean]
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 547831
Target Species Plant
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.