missing translation for 'onlineSavingsMsg'
Learn More

Melford Trypsin Inhibitor

Product Code. 30018835
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30018835 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30018835

Brand: Melford T230251.0

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Serine protease inhibitor specific for trypsin and trypsin like proteases.

Serine protease inhibitor specific for trypsin and trypsin like proteases.

Specifications

CAS 9035-81-8
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.