missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Trypsin-EDTA (0.05%)
Enzymatic cell dissociation reagent
Brand: STEMCELL Technologies 07910
Additional Details : Weight : 0.50000kg
Description
- Trypsin-EDTA (0.05%) can be used to achieve a single-cell suspension of human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells for flow cytometry or cloning assays
- This is a gentler version than the standard concentration (0.25%), and has applications to many other cell culture systems in addition to human ES cells and iPS cells
Specifications
500 mL |