missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Trypsin (Bovine), Modified, Sequencing Grade

Product Code. 16151840
Click to view available options
Quantity:
4 x 25 μg
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 16151840

Brand: Abnova™ P5320.4x25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

Accession Number Q29463
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 282603/615237
Name Trypsin (Bovine), Modified, Sequencing Grade
Preparation Method Native protein purified from Bovine Pancreas. Further chemically modified to promote stability and purified to remove autolysis fragments, resulting in a highly stable trypsin product resistant to autolysis while retaining specificity.
Quantity 4 x 25 μg
Storage Requirements Store at -20°C on dry atmosphere.
Regulatory Status RUO
Gene Alias TRYP8
Common Name PRSS2
Gene Symbol PRSS2
Biological Activity ≥4 units per mg protein. One Unit is equivalent to one micromole of TCA soluble products, measured as tyrosine, released from 2% casein per minute, in 0.05M Tris-HCl, pH 7.6, at 37°C, in a 30 minute reaction.
Species Bovine
Protein Tag None
Expression System Native protein purified from Bovine Pancreas. Further chemically modified to promote stability and purified to remove autolysis fragments, resulting in a highly stable trypsin product resistant to autolysis while retaining specificity.
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.