missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trypsin 3/PRSS3 Antibody (420025) [Janelia Fluor™ 549], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3710I
This item is not returnable.
View return policy
Description
Trypsin 3/PRSS3 Monoclonal antibody specifically detects Trypsin 3/PRSS3 in Human samples. It is validated for Neutralization
Specifications
| Trypsin 3/PRSS3 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Brain trypsinogen, EC 3.4.21, EC 3.4.21.4, Mesotrypsinogen, pancreatic trypsinogen III, protease, serine, 3, protease, serine, 3 (mesotrypsin), protease, serine, 4 (trypsin 4, brain), PRSS4, Serine protease 3, Serine protease 4, T9, TRY3MTG, TRY4mesotrypsin, trypsin 3, Trypsin III, Trypsin IV, trypsin-3, trypsinogen 4, trypsinogen 5, trypsinogen IV | |
| Mouse myeloma cell line NS0-derived recombinant human Trypsin 3/PRSS3, Val16-Ser247, Accession # NP_002762 | |
| 0.1 mL | |
| Cancer, Proteases & Other Enzymes | |
| 5646 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Neutralization | |
| 420025 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction