missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Trypsin 2 Recombinant Rabbit Monoclonal Antibody (3D9O7)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA537902

Product Code. 17206783

  • £463.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Samples: Mouse pancreas, Rat pancreas Immunogen sequence: MNLLLILTFV AAAVAAPFDD DDKIVGGYIC EENSVPYQVS LNSGYHFCGG SLISEQWVVS AGHCYKSRIQ VRLGEHNIEV LEGNEQFINA AKIIRHPKYN.

This gene belongs to the trypsin family of serine proteases and encodes anionic trypsinogen. It is part of a cluster of trypsinogen genes that are located within the T cell receptor beta locus. Enzymes of this family cleave peptide bonds that follow lysine or arginine residues. This protein is found at high levels in pancreatic juice and its upregulation is a characteristic feature of pancreatitis. This protein has also been found to activate pro-urokinase in ovarian tumors, suggesting a function in tumor invasion. In addition, this enzyme is able to cleave across the type II collagen triple helix in rheumatoid arthritis synovitis tissue, potentially participating in the degradation of type II collagen-rich cartilage matrix. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Trypsin 2
Recombinant Monoclonal
0.33 mg/mL
PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
P00763, P07146, P07478
PRSS2
Synthetic peptide.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot
3D9O7
Unconjugated
PRSS2
anionic trypsin; anionic trypsin II; Anionic trypsin-2; anionic trypsinogen; EC 3.4.21; EC 3.4.21.4; MGC111183; MGC120174; MGC120175; pancreatic anionic trypsinogen; pancreatic trypsin 2; pancreatic trypsin II; pretrypsinogen II; protease serine 2 preproprotein; protease, serine 2; protease, serine, 2; protease, serine, 2 (trypsin 2); PRSS2; Ptryss2; Serine protease 2; Ta; Tesp4; Try2; TRY8; TRYP; TRYP2; TRYP8; trypsin 2; trypsin II; trypsin Trypsin II; Trypsin-2; trypsinogen 2; trypsinogen 20; trypsinogen anionic precursor
Rabbit
Affinity chromatography
RUO
22072, 25052, 5645
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.