missing translation for 'onlineSavingsMsg'
Learn More

Trypsin 2/PRSS2 Antibody (410322) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30731176 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731176 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731176 Supplier Novus Biologicals Supplier No. FAB3586J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Trypsin 2/PRSS2 Monoclonal antibody specifically detects Trypsin 2/PRSS2 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trypsin 2/PRSS2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 410322
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Anionic trypsinogen, EC 3.4.21, EC 3.4.21.4, MGC111183, MGC120174, MGC120175, protease serine 2 preproprotein, protease, serine, 2 (trypsin 2), protease, serine, 2, preproprotein, Serine protease 2, TRY2TRY8, TRYP2, trypsin 2, Trypsin II, trypsin-2, trypsinogen 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Trypsin 2/PRSS2, Ala16-Ser247, Accession # P07478
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 5645
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.