missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trypsin 1/PRSS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43272
This item is not returnable.
View return policy
Description
Trypsin 1/PRSS1 Polyclonal antibody specifically detects Trypsin 1/PRSS1 in Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Trypsin 1/PRSS1 | |
| Polyclonal | |
| Unconjugated | |
| 0.01 M PBS, pH 7.4, 50% glycerol | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 0.5 mg/mL | |
| Western Blot 0.5-2ug/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation | |
| Beta-trypsin, Cationic trypsinogen, digestive zymogen, DKFZp779A0837, EC 3.4.21, EC 3.4.21.4, FLJ57296, MGC120175, MGC149362, nonfunctional trypsin 1, protease, serine, 1 (trypsin 1), Serine protease 1, TRP1, TRY1beta-trypsin, TRY4, TRYP1, Trypsin I, trypsin-1, trypsinogen 1, trypsinogen A | |
| Immunized with recombinant Mouse Trypsin 1/PRSS1 (Uniprot#: Q9Z1R9; Lys23∽Asn246) | |
| 100 μL | |
| Cancer | |
| 5644 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction