missing translation for 'onlineSavingsMsg'
Learn More

Trypan Blue (Technical Grade), TRC

Product Code. 30135379
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135379 10 g 10g
30006759 5 g 5g
30010148 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135379 Supplier Toronto Research Chemicals Supplier No. TRCT89552310G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-57-1
Molecular Formula C34 H24 N6 O14 S4 . 4 Na
Molecular Weight (g/mol) 960.8
Synonym C.I. Direct Blue 14, Tetrasodium Salt, Diphenyl Blue, Amanil Sky Blue R, Bencidal Blue 3B, Benzamine Blue, Benzanil Blue 3BN, Benzo Blue 3BS, Benzo Blue, Blue 3B, Blue EMB, Brasilamina Blue 3B, C.I. 23850, C.I. Direct Blue 14, Chloramine Blue 3B, Chrome Leather Blue 3B, Congo Blue, Cresotine Blue 3B, Diamine Blue 3B, Diamineblue, Diaphtamine Blue TH, Diazine Blue 3B, Diazol Blue 3B, Diphenyl Blue 3B, Direct Blue 14, Direct Blue 3B, Direct Blue M 3B, Hispamin Blue 3BX, Naphthylamine Blue, Niagara Blue, Niagara Blue 3B, Paramine Blue 3B, Pontamine Blue 3BX, Sodium Ditolyldisazobis-8-amino-1-naphthol-3,6-disulfonate, Trypan (Congo) Blue, Trypan Blue BPC, Trypane Blue, 3,3'-[(3,3'-Dimethyl[1,1'-biphenyl]-4,4'-diyl)bis(azo)]bis[5-amino-4-hydroxy-2,7-naphthalenedisulfonic Acid Tetrasodium Salt
IUPAC Name tetrasodium;5-amino-3-[(E)-[4-[4-[(E)-(8-amino-1-hydroxy-3,6-disulfonato-2-naphthyl)azo]-3-methyl-phenyl]-2-methyl-phenyl]azo]-4-hydroxy-naphthalene-2,7-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].Cc1cc(ccc1N=Nc2c(O)c3c(N)cc(cc3cc2S(=O)(=O)[O-])S(=O)(=O)[O-])c4ccc(N=Nc5c(O)c6c(N)cc(cc6cc5S(=O)(=O)[O-])S(=O)(=O)[O-])c(C)c4

Specifications

Quantity 10 g
InChI Formula InChI=1S/C34H28N6O14S4.4Na/c1-15-7-17(3-5-25(15)37-39-31-27(57(49,50)51)11-19-9-21(55(43,44)45)13-23(35)29(19)33(31)41)18-4-6-26(16(2)8-18)38-40-32-28(58(52,53)54)12-20-10-22(56(46,47)48)14-24(36)30(20)34(32)42;;;;/h3-14,41-42H,35-36H2,1-2H3,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4/b39-37+,40-38+;;;;
Formula Weight 959.9824
Chemical Name or Material Trypan Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.