missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >280.0°C |
| Dye Content | 60% min. |
| Solubility Information | Solubility in water: 10g/L (25°C). Other solubilities: practically insoluble in alcohol, soluble in cellosolve, ethylene glycol, acids |
| Infrared Spectrum | Authentic |
| IUPAC Name | tetrasodium;(3Z)-5-amino-3-[[4-[4-[(2Z)-2-(8-amino-1-oxo-3,6-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methylphenyl]-2-methylphenyl]hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate |
| Formula Weight | 960.78 |
| Packaging | Glass bottle |
| UV Spectrometry | Complies |
| Color | Black/Brown to Blue |
| Quantity | 100 g |
| Show More |
Safety and Handling
Product Identifier
- Trypan Blue
- Danger
- Carcinogenicity Category 1B
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P233-Keep container tightly closed.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?