missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TrueTag™ Donor DNA Kit, GFP

Product Code. 17103209
Change view
Click to view available options
Quantity:
10 Reactions
Unit Size:
10 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17103209 10 Reactions 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price
Product Code. 17103209 Supplier Invitrogen™ Supplier No. A42992

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price

The Invitrogen™ TrueTag™ Donor DNA Kit, GFP, provides a rapid solution to drive high-efficiency gene tagging (up to 100% edited cells) with transfection-ready donor DNA for genome editing experiments.

The Invitrogen TrueTag Donor DNA Kit, GFP, provides a rapid solution to drive high-efficiency gene tagging (up to 100% edited cells) with transfection-ready donor DNA for genome editing experiments.

Benefits of the TrueTag Donor DNA Kit include:
• One-step PCR amplification to generate donor templates—no cloning required
• Generate transfection-ready DNA donors in hours instead of days
• Enrich up to 100% GFP-tagged cells

The TrueTag Donor DNA Kit, GFP, provides everything you need to produce a high-quality DNA donor to tag your gene with a selection marker and GFP. Simply provide two locus-specific primers to add homology arms, PCR-amplify, and purify for a transfection-ready donor in hours, not days. Each kit is designed to enable you to:
• Tag the carboxy- (C-) or amino- (N-) terminus of your gene of interest
• Choose from puromycin or blasticidin selection markers—whatever best suits your cell model
• Easily remove antibiotic resistance markers with Cre/Lox following cell enrichment

Included in the kit
• Four linear donor templates: N-terminus/puromycin resistance, N-terminus/blasticidin resistance, C-terminus/puromycin resistance, C-terminus/blasticidin resistance
• Phusion Flash High-Fidelity PCR Master Mix
• PCR cleanup columns and buffers
• Positive control primers to tag human beta-actin (ACTB)

Intuitive software to rapidly design gene-specific reagents
Target-specific gene editing reagents are required for proper knock-in of your fluorescent tag. The Invitrogen TrueDesign Genome Editor web-based software was specifically created to:
1. Identify your target gene transcript
2. Designate your insertion site
3. Design and enable easy ordering of all necessary components

Required reagents available via TrueDesign software (not included in kit)
• CRISPR guide RNA or TALENs designed adjacent to N- or C-terminus
• Oligos to PCR-amplify your donor and add appropriate homology arms
• Sequencing primers for assessment of CRISPR-Cas9 cleavage efficiency
• Verification primers to validate proper insertion of the TrueTag DNA Donor

Also required and available for purchase separately or as part of TrueDesign workflow:
• TrueCut Cas9 Protein v2, TrueCut HiFi Cas9 Protein, or GeneART CRISPR mRNA
• CRISPRMAX Transfection Reagent or MessengerMAX Transfection Reagent

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • TrueTag Assembly Kit, store at -20°C
• TrueTag Cleanup Kit, store at room temperature

Description Tag: GFP
Format Kit
For Use With (Application) Fluorescent gene tagging, cell line development
Label or Dye GFP
Product Line TrueTag
Product Type Donor DNA Kit
Quantity 10 Reactions
Shipping Condition Approved for shipment on Wet or Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.