missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TrueGuide™ sgRNA Positive Control, Rosa26 (mouse)

Product Code. 31662353
Change view
Click to view available options
Quantity:
3 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31662353 3 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31662353 Supplier Invitrogen™ Supplier No. A35525
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for the optimization of transfection and verification of editing efficiency in transfected cells.

Provides complete collection of validated positive controls for the TrueGuide Synthetic gRNA product line to help maximize editing performance. This positive control against ROSA26 targets intronic region of gene and has achieved editing efficiencies of up to 80% in variety of mouse cell lines. ROSA26 control easily determines the optimal conditions for delivering CRISPR/Cas9 tools to cell type, allowing to move forward in editing experiments and minimize downstream effort to isolate and validate edited clones

Validated positive control crRNA targeting Rosa26

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Delivered as dried RNA. Store dried RNA at room temperature; store resuspended RNA at -5°C to -30°C. Resuspend in nuclease-free water or TE.
Format Tube
Control Type Positive Control
RNAi Type gRNA
Species Mouse
Product Line TrueGuide
Product Type gRNA
Quantity 3 nmol
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.