missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TrueGuide™ sgRNA Negative Control, non-targeting 1

Product Code. 31662581
Change view
Click to view available options
Quantity:
3 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31662581 3 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31662581 Supplier Invitrogen™ Supplier No. A35526
  • ONLINE EXCLUSIVE PRICE
    £196.00
    £189.65 / Pack of 1
    Save £6.35
    3% Off
View all promotions
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TrueGuide Synthetic sgRNA Negative Control does not recognize any sequence in the human or mouse genome and provides a non-edited experimental control.

TrueGuide negative controls are non-targeting gRNA sequences that do not recognize any sequence in the human or mouse genome and provide a non-edited control when performing analysis on pooled populations of edited cells.

Non-targeting gRNA sequences that do not recognize any sequence in the human or mouse genome and provide a non-edited control when performing analysis on pooled populations of edited cells.

Validated negative control crRNA. Universal control for Mouse and Human

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Delivered as dried RNA. Store dried RNA at room temperature; store resuspended RNA at -5°C to -30°C. Resuspend in nuclease-free water or TE.
Format Tube
Control Type Negative Control
RNAi Type gRNA
Species Human, Mouse
Product Line TrueGuide
Product Type gRNA
Quantity 3 nmol
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.