missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TrueCut™ HiFi Cas9 Protein (1 μg/μL)

Product Code. 17474673
Change view
Click to view available options
Quantity:
10 μg
25 μg
Concentration:
1 μg/μL
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Concentration unitSize
17474673 10 μg 1 μg/μL 10µg
17484673 25 μg 1 μg/μL 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price
Product Code. 17474673 Supplier Invitrogen™ Supplier No. A50574

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price

A high fidelity CRISPR Cas9 protein variant engineered to demonstrate superior off-target profiles, while preserving maximum on-target editing efficiency. TrueCut™ HiFi Cas9 Protein is ideal for applications that are sensitive to off-target events and when more precise editing is required.

  • A high fidelity CRISPR Cas9 protein variant engineered to demonstrate superior off-target profiles, while preserving maximum on-target editing efficiency
  • Near complete elimination of off-target events in all tested cell lines including standard, immune, primary, and stem
  • Maintains maximum on-target editing efficiency compared to TrueCut™ Cas9 Protein v2
  • Transfection-ready using either electroporation or lipid-mediated transfection reagents
  • Manufactured under strict ISO-13485 quality standards
  • Ideal for applications that are sensitive to off-target events and when more precise editing is required
  • Eliminates time consuming cloning steps

Available in Two Concentrations

  • 1 μg/μL - For use in standard editing scenarios
  • 5 μg/μL - For optimized editing conditions in more difficult scenarios such as in primary or embryonic cells, microinjection, or when screening multiple gRNA sequences at a time
  • For custom sizes or concentrations, please inquire
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 μg/μL
Purity ≥90%
Sensitivity Yes
Source E. coli
Quantity 10 μg
Product Type HiFi Cas9
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.