missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ True North™ Mini-Cooler

Product Code. 16330686 Shop All Thermo Scientific CO₂ Incubators Products
Click to view available options
Array:
3 x 4
4 x 8
8 x 12
Color:
Blue
Green
Description:
Clear Lid
Gel Filled Lid
Withstand Temperature (Metric):
-20°C
0°C
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16330686

Brand: Heathrow Scientific™ HS120074

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The True North™ Mini Cooler protects temperature-sensitive samples at the bench or in the freezer. The Mini Cooler is ideal for keeping PCR™ reagents, enzymes, and other biochemical reagents cool while working at the bench.

  • Polycarbonate construction
  • Cooler has rubber non-slip feet
  • Gridded and keyed one-position lid for easy sample identification
  • Available with lid filled with insulating gel

Specifications

Color Blue
Height (Metric) 10 cm
Height (English) 3.9 in.
Length (Metric) 24.5 cm
Length (English) 9.6 in.
Width (Metric) 10.5 cm
Width (English) 4.1 in.
Description Clear Lid
For Use With (Application) Tubes
Holds Tubes
Quantity 1 Ea.
Array 4 x 8
Material Polycarbonate
Type Cooler
Capacity (Metric) 0.5 to 2 mL
Withstand Temperature (Metric) -20°C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.