missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ True North™ Mini-Cooler

Product Code. 16310686 Shop All Thermo Scientific CO₂ Incubators Products
Change view
Click to view available options
Array:
3 x 4
4 x 8
8 x 12
Color:
Blue
Green
Description:
Clear Lid
Gel Filled Lid
Withstand Temperature (Metric):
-20°C
0°C
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Array Color Description Withstand Temperature (Metric) unitSize
16310686 4 x 8 Green Gel Filled Lid 0°C Each
16390676 3 x 4 Green Clear Lid 0°C Each
16300686 4 x 8 Green Clear Lid 0°C Each
16320686 3 x 4 Blue Clear Lid -20°C Each
16330686 4 x 8 Blue Clear Lid -20°C Each
16340686 4 x 8 Blue Gel Filled Lid -20°C Each
16350686 8 x 12 Blue Clear Lid -20°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16310686 Supplier Heathrow Scientific™ Supplier No. HS120072
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The True North™ Mini Cooler protects temperature-sensitive samples at the bench or in the freezer. The Mini Cooler is ideal for keeping PCR™ reagents, enzymes, and other biochemical reagents cool while working at the bench.

  • Polycarbonate construction
  • Cooler has rubber non-slip feet
  • Gridded and keyed one-position lid for easy sample identification
  • Available with lid filled with insulating gel

Specifications

Color Green
Height (Metric) 10 cm
Height (English) 3.9 in.
Length (Metric) 24.5 cm
Length (English) 9.6 in.
Width (Metric) 10.5 cm
Width (English) 4.1 in.
Description Gel Filled Lid
For Use With (Application) Tubes
Holds Tubes
Quantity 1 Ea.
Array 4 x 8
Material Polycarbonate
Type Cooler
Capacity (Metric) 0.5 to 2 mL
Withstand Temperature (Metric) 0°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.