missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRUB2 Recombinant Rabbit Monoclonal Antibody (K01_2J62)
Description
TRUB2 Recombinant Monoclonal Antibody for Western Blot
Pseudouridine is an abundant component of rRNAs and tRNAs and is enzymatically generated by isomerization of uridine by pseudouridine synthase (Zucchini et al., 2003 [PubMed 12736709]).
Specifications
Specifications
| Antigen | TRUB2 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2J62 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | TRUB2 |
| Gene Accession No. | O95900 |
| Gene Alias | CLONE24922; G430055L02Rik; Mitochondrial mRNA pseudouridine synthase TRUB2; probable tRNA pseudouridine synthase 2; RP11-339B21.1; TruB pseudouridine (psi) synthase family member 2; TruB pseudouridine (psi) synthase homolog 2; TruB pseudouridine (psi) synthase homolog 2 (E. coli); TruB pseudouridine synthase family member 2; Trub2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?