missing translation for 'onlineSavingsMsg'
Learn More

TRUB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18365634 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365634 100 μg 100µL
18354534 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18365634 Supplier Novus Biologicals Supplier No. NBP316027100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRUB1 Polyclonal antibody specifically detects TRUB1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRUB1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias EC 5.4.99, EC 5.4.99.-, PUS4probable tRNA pseudouridine synthase 1, TruB pseudouridine (psi) synthase homolog 1 (E. coli)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-280 of human TRUB1 (NP_631908.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142940
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.