missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRSPAP1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94804-0.1ml
This item is not returnable.
View return policy
Description
TRSPAP1 Polyclonal antibody specifically detects TRSPAP1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TRSPAP1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| FLJ20503, PRO1902, SECp43, SECP43RP4-669K10.4, tRNA selenocysteine 1 associated protein 1, tRNA selenocysteine 1-associated protein 1, tRNA selenocysteine associated protein (SECP43), tRNA selenocysteine associated protein 1, tRNA selenocysteine-associated protein 1, TRSPAP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-287 of human TRNAU1AP (NP_060316.1). MAASLWMGDLEPYMDENFISRAFATMGETVMSVKIIRNRLTGIPAGYCFVEFADLATAEKCLHKINGKPLPGATPAKRFKLNYATYGKQPDNSPEYSLFVGDLTPDVDDGMLYEFFVKVYPSCRGGKVVLDQTGVSKGYGFVKFTDELEQKRALTECQGAVGLGSKPVRLSVAIPKASRVKPVEYSQMYSYSYNQYYQQYQNYYAQWGYDQNTGSYSYSYPQYGYTQSTMQTYEEVGDDALEDPMPQLDVTEANKEFMEQSEELYDALMDCHWQPLDTVSSEIPAMM | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54952 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction