missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRRAP Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-58810FR
This item is not returnable.
View return policy
Description
TRRAP Polyclonal antibody specifically detects TRRAP in Human samples. It is validated for Western Blot
Specifications
| TRRAP | |
| Polyclonal | |
| Western Blot | |
| 350/400 kDa PCAF-associated factor, FLJ10671, PAF350/400, PAF400STAF40, Tra1, Tra1 homolog, transformation/transcription domain-associated protein, TR-AP | |
| The immunogen recognized by this antibody maps to a region between residue 3275 and 3325 of human transformation/transcription domain-associated protein using the numbering given in entry AAD04629.1 (GeneID 8295). | |
| 0.1 mL | |
| Cell Cycle and Replication, Protein Kinase | |
| 8295 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction