missing translation for 'onlineSavingsMsg'
Learn More

TRRAP Antibody [DyLight 680], Novus Biologicals™

Product Code. 30732153 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732153 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732153 Supplier Novus Biologicals Supplier No. AF7365Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

TRRAP Polyclonal antibody specifically detects TRRAP in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRRAP
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias 350/400 kDa PCAF-associated factor, FLJ10671, PAF350/400, PAF400STAF40, Tra1, Tra1 homolog, transformation/transcription domain-associated protein, TR-AP
Host Species Sheep
Immunogen E. coli-derived recombinant human TRRAP, Asn3610-Leu3859, predicted, Accession # Q9Y4A5
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 8295
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.