missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPV5 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-93520
This item is not returnable.
View return policy
Description
TRPV5 Polyclonal specifically detects TRPV5 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| TRPV5 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Calcium transport protein 2, calcium transporter 2, ECaC, ECaC1, ECAC1CaT2, Epithelial calcium channel 1CAT2, Osm-9-like TRP channel 3, OTRPC3, transient receptor potential cation channel subfamily V member 5, transient receptor potential cation channel, subfamily V, member 5, TrpV5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 56302 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/mL, Peptide ELISA Detection limit 1:16000 | |
| NP_062815.2 | |
| TRPV5 | |
| Peptide with sequence C-SHRGWEILRQNT corresponding to internal region according to NP_062815.2. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, Pig and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction