missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPV5 Antibody (6D6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00056302-M06
This item is not returnable.
View return policy
Description
TRPV5 Monoclonal antibody specifically detects TRPV5 in Human samples. It is validated for Western Blot, ELISA
Specifications
| TRPV5 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Calcium transport protein 2, calcium transporter 2, ECaC, ECaC1, ECAC1CaT2, Epithelial calcium channel 1CAT2, Osm-9-like TRP channel 3, OTRPC3, transient receptor potential cation channel subfamily V member 5, transient receptor potential cation channel, subfamily V, member 5, TrpV5 | |
| TRPV5 (NP_062815, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGGFLPKAEGPGSQLQKLLPSFLVREQDWDQHLDKLHMLQQKRILESPLLRASKENDLSVLRQLLLDCTCDVRQRGALGETALHIAALYDNLEAALVLME | |
| 0.1 mg | |
| Cancer, Endocrinology, Signal Transduction | |
| 56302 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 6D6 | |
| Western Blot 1:500, ELISA | |
| NP_062815 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction