missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPV4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03968-100UL
This item is not returnable.
View return policy
Description
TRPV4 Polyclonal specifically detects TRPV4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TRPV4 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 59341 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| CMT2Cosmosensitive transient receptor potential channel 4, HMSN2C, Osm-9-like TRP channel 4, OTRPC4SSQTL1, transient receptor potential cation channel subfamily V member 4, transient receptor potential cation channel, subfamily V, member 4, Transient receptor potential protein 12, TRP12OSM9-like transient receptor potential channel 4, TrpV4, Vanilloid receptor-like channel 2, Vanilloid receptor-like protein 2, vanilloid receptor-related osmotically activated channel, Vanilloid receptor-related osmotically-activated channel, VRL2, VRL-2, VR-OAC, VROACSPSMA | |
| A synthetic peptide corresponding to a sequence within amino acids 800 to the C-terminus of human TRPV4 (NP_067638.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction