Learn More
TRPV4 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5111758
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)EDQSN(S )TVPSYPA(S )RD Homology: Rat - 14/16 amino acid residues identical; mouse - 13/16 amino acid residues identical; human - 12/16 amino acid residues identical.
TRPV4 (Transient Receptor Potential Cation Channel Subfamily V Member 4) is a cation-selective channel, activated in response to systemic osmotic pressure. Non-selective calcium permeant cation channel probably involved in osmotic sensitivity and mechanosensitivity. TRPV4 is a member of the OSM9-like transient receptor potential channel (OTRPC) subfamily in the transient receptor potential (TRP) superfamily of ion channels. The TRPV4 protein is a Ca2+-permeable, nonselective cation channel that is thought to be involved in the regulation of systemic osmotic pressure. Mutations in the TRPV4 gene are the cause of spondylometaphyseal and metatropic dysplasia and hereditary motor and sensory neuropathy type IIC. Diseases associated with TRPV4 include Metatropic Dysplasia and Spondylometaphyseal Dysplasia, Kozlowski Type. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| TRPV4 (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| Trpv4 | |
| 0610033B08Rik; BCYM3; BOS_16634; CMT2C; HMSN2C; Osm-9-like TRP channel 4; OTRPC4; SMAL; SPSMA; SSQTL1; Transient receptor potential cation channel subfamily V member 4; Transient receptor potential protein 12; Trp12; Trpv4; Vanilloi; Vanilloid receptor-like channel 2; Vanilloid receptor-like protein 2; Vanilloid receptor-related osmotically-activated channel; Vrl2; VRL-2; Vroac; VR-OAC; wu:fp52e02 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 59341, 66026 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunomicroscopy, Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| Q9ERZ8, Q9HBA0 | |
| Trpv4 | |
| Peptide corresponding to amino acid residues 647-662 of rat TRPV4 (3rd extracellular loop) | |
| 50 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.