missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRPV3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599875
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Antibody detects endogenous levels of total TRPV3.
TRPV3 belongs to a family of nonselective cation channels that function in a variety of processes, including temperature sensation and vasoregulation. The thermosensitive members of this family are expressed in subsets of sensory neurons that terminate in the skin, and are activated at distinct physiological temperatures. This channel is activated at temperatures between 22 and 40 degrees C. This gene lies in close proximity to another family member (TRPV1) gene on chromosome 17, and the two encoded proteins are thought to associate with each other to form heteromeric channels.
Tekniske data
| TRPV3 | |
| Polyclonal | |
| Unconjugated | |
| TRPV3 | |
| 1110036I10Rik; AI644701; FNEPPK2; heat sensitive channel TRPV3; MGC124324; MGC124325; Nh; non-hair; OLMS; Transient receptor potential cation channel subfamily V member 3; transient receptor potential cation channel, subfamily V, member 3; TRP ion channel TRPV3; TrpV3; Vanilloid receptor-like 3; vanilloid receptor-related osmotically activated channel protein; VRL 3; VRL3; VRL-3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 162514, 246788 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8K424, Q8NET8 | |
| TRPV3 | |
| A synthesized peptide derived from human TRPV3(Accession Q8NET8), corresponding to amino acid residues G21-N71. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion