missing translation for 'onlineSavingsMsg'
Learn More

TRPS1 Rabbit Polyclonal Antibody, Proteintech

Product Code. p-200107709 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30234951 150 μL 150µL
30236548 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30234951 Supplier Proteintech Supplier No. 287531AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The TRPS1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRPS1. This antibody recognizes human antigen. The TRPS1 antibody has been validated for the following applications: WB, IHC, ELISA analysis.

Specifications

Antigen TRPS1
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Concentration 600 μg/mL
Conjugate Unconjugated
Gene Symbols GC79; LGCR; MGC134928
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Antigen affinity purification
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7227
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 142 kDa
Product Type Polyclonal Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.