missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRPS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549890
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total TRPS1 protein.
This gene encodes a transcription factor that represses GATA-regulated genes and binds to a dynein light chain protein. Binding of the encoded protein to the dynein light chain protein affects binding to GATA consensus sequences and suppresses its transcriptional activity. Defects in this gene are a cause of tricho-rhino-phalangeal syndrome (TRPS) types I-III. [provided by RefSeq].
Specifications
| TRPS1 | |
| Polyclonal | |
| Unconjugated | |
| TRPS1 | |
| AI115454; AI447310; D15Ertd586e; GC79; LGCR; transcriptional repressor GATA binding 1; trichorhinophalangeal syndrome I; trichorhinophalangeal syndrome I (human); trichorhinophalangeal syndrome I homolog; tricho-rhino-phalangeal syndrome type I protein; TRPS1; Zinc finger protein GC79; zinc finger transcription factor Trps1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7227, 83925 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q925H1, Q9UHF7 | |
| TRPS1 | |
| Synthesized peptide derived from N-terminal of human TRPS1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction