Learn More
TRPS1 Mouse, anti-Human, Clone: 1B1G2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68562-1-IG-20UL
This item is not returnable.
View return policy
Description
The TRPS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRPS1. This antibody recognizes human antigen. The TRPS1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TRPS1 | |
| 1B1G2 | |
| GC79, LGCR, TRPS1, Zinc finger protein GC79 | |
| 20 μL | |
| 7227 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |