missing translation for 'onlineSavingsMsg'
Learn More

TRPS1 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18420548 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420548 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420548 Supplier Novus Biologicals Supplier No. NBP204120DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRPS1 Polyclonal antibody specifically detects TRPS1 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPS1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias trichorhinophalangeal syndrome I, tricho-rhino-phalangeal syndrome type I protein
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1000 and 1050 of human Tricho-Rhino-Phalangeal Syndrome Type I using the numbering given in entry AAG21134.1 (GeneID 7227).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7227
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.