missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94139-0.1ml
This item is not returnable.
View return policy
Description
TRPM8 Polyclonal antibody specifically detects TRPM8 in Human samples. It is validated for Western Blot
Specifications
| TRPM8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Long transient receptor potential channel 6, LTrpC-6, LTRPC6, MGC2849, short form of the TRPM8 cationic channel, transient receptor potential cation channel, subfamily M, member 8, Transient receptor potential p8, transient receptor potential subfamily M member 8, trp-p8, TRPP8transient receptor potential cation channel subfamily M member 8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 980-1104 of human TRPM8 (NP_076985.4). GYTVGTVQENNDQVWKFQRYFLVQEYCSRLNIPFPFIVFAYFYMVVKKCFKCCCKEKNMESSVCCFKNEDNETLAWEGVMKENYLVKINTKANDTSEEMRHRFRQLDTKLNDLKGLLKEIANKIK | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 79054 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction