missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM7 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | TRPM7 |
|---|---|
| Concentration | 700 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234100
|
Proteintech
55251-1-AP-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233237
|
Proteintech
55251-1-AP-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
The TRPM7 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human TRPM7. This antibody recognizes human, mouse, rat antigen. The TRPM7 antibody has been validated for the following applications: WB, ELISA analysis.Specifikationer
| TRPM7 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| CHAK; CHAK1; FLJ20117; FLJ25718; LTRPC7; TRP-PLIK | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 700 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 54822 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel