missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRPM7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587618
This item is not returnable.
View return policy
Description
Immunogen sequence: NSWYKVILSI LVPPAILLLE YKTKAEMSHI PQSQDAHQMT MDDSENNFQN ITEEIPMEVF KEVRILDSNE GKNEMEIQMK SKKLPITRKF YAFYHAPIVK; Positive Samples: Mouse skeletal muscle; Cellular Location: Membrane, Multi-pass membrane protein.
TRPCs, mammalian homologs of the Drosophila transient receptor potential (trp) protein, are ion channels that are thought to mediate capacitative calcium entry into the cell. TRP-PLIK is a protein that is both an ion channel and a kinase. As a channel, it conducts calcium and monovalent cations to depolarize cells and increase intracellular calcium. As a kinase, it is capable of phosphorylating itself and other substrates. The kinase activity is necessary for channel function, as shown by its dependence on intracellular ATP and by the kinase mutants.
Specifications
| TRPM7 | |
| Polyclonal | |
| Unconjugated | |
| Trpm7 | |
| 2310022G15Rik; 4833414K03Rik; 5033407O22Rik; ALSPDC; CHAK; CHAK1; Channel kinase 1; Channel-kinase 1; FLJ20117; long transient receptor potential channel 7; long transient receptor potential channel 7 {ECO:0000250; Ltpr7; LTRPC ion channel family member 7; LTRPC7; LTrpC-7; LTrpC7 {ECO:0000250; PubMed:11161216}; RGD:620053}; tr; transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel subfamily M member 7 {ECO:0000312; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential M7; transient receptor potential-phospholipase C-interacting kinase; transient receptor potential-phospholipase C-interacting kinase {ECO:0000303; transient receptor potential-related protein, ChaK; TRMP7; TRP PLIK; Trpm7; trpm7 {ECO:0000312; TRPPLIK; Trp-plik; TRP-PLIK {ECO:0000303; UniProtKB:Q96QT4} | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 58800 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.96 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q923J1 | |
| Trpm7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 756-855 of human TRPM7 (NP_0601423). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction