Learn More
Invitrogen™ TRPM7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595588
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human COLO-320 whole cell, human 22RV1 whole cell, human SGC-7901 whole cell. ICC/IF: MCF7 cell. Flow: THP-1 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
TRPCs, mammalian homologs of the Drosophila transient receptor potential (trp) protein, are ion channels that are thought to mediate capacitative calcium entry into the cell. TRP-PLIK is a protein that is both an ion channel and a kinase. As a channel, it conducts calcium and monovalent cations to depolarize cells and increase intracellular calcium. As a kinase, it is capable of phosphorylating itself and other substrates. The kinase activity is necessary for channel function, as shown by its dependence on intracellular ATP and by the kinase mutants.
Specifications
| TRPM7 | |
| Polyclonal | |
| Unconjugated | |
| Trpm7 | |
| 2310022G15Rik; 4833414K03Rik; 5033407O22Rik; ALSPDC; CHAK; CHAK1; Channel kinase 1; Channel-kinase 1; FLJ20117; long transient receptor potential channel 7; long transient receptor potential channel 7 {ECO:0000250; Ltpr7; LTRPC ion channel family member 7; LTRPC7; LTrpC-7; LTrpC7 {ECO:0000250; PubMed:11161216}; RGD:620053}; tr; transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel subfamily M member 7 {ECO:0000312; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential M7; transient receptor potential-phospholipase C-interacting kinase; transient receptor potential-phospholipase C-interacting kinase {ECO:0000303; transient receptor potential-related protein, ChaK; TRMP7; TRP PLIK; Trpm7; trpm7 {ECO:0000312; TRPPLIK; Trp-plik; TRP-PLIK {ECO:0000303; UniProtKB:Q96QT4} | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54822, 58800, 679906 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q923J1, Q925B3, Q96QT4 | |
| Trpm7 | |
| E.coli-derived human TRPM7 recombinant protein (Position: K777-K905). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.