missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRPM7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102835
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TRPM7.
TRPCs, mammalian homologs of the Drosophila transient receptor potential (trp) protein, are ion channels that are thought to mediate capacitative calcium entry into the cell. TRP-PLIK is a protein that is both an ion channel and a kinase. As a channel, it conducts calcium and monovalent cations to depolarize cells and increase intracellular calcium. As a kinase, it is capable of phosphorylating itself and other substrates. The kinase activity is necessary for channel function, as shown by its dependence on intracellular ATP and by the kinase mutants.
Specifications
| TRPM7 | |
| Polyclonal | |
| Unconjugated | |
| Trpm7 | |
| 2310022G15Rik; 4833414K03Rik; 5033407O22Rik; ALSPDC; CHAK; CHAK1; Channel kinase 1; Channel-kinase 1; FLJ20117; long transient receptor potential channel 7; long transient receptor potential channel 7 {ECO:0000250; Ltpr7; LTRPC ion channel family member 7; LTRPC7; LTrpC-7; LTrpC7 {ECO:0000250; PubMed:11161216}; RGD:620053}; tr; transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel subfamily M member 7 {ECO:0000312; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential M7; transient receptor potential-phospholipase C-interacting kinase; transient receptor potential-phospholipase C-interacting kinase {ECO:0000303; transient receptor potential-related protein, ChaK; TRMP7; TRP PLIK; Trpm7; trpm7 {ECO:0000312; TRPPLIK; Trp-plik; TRP-PLIK {ECO:0000303; UniProtKB:Q96QT4} | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54822, 58800, 679906 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q923J1, Q925B3, Q96QT4 | |
| Trpm7 | |
| A synthesized peptide derived from human TRPM7(Accession Q96QT4), corresponding to amino acid residues D1832-L1865. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction